To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Serine/Threonine Kinase 2
- Antigen Symbol:STK2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:STK2|SLK|Serine/threonine protein kinase 2|CTCL tumor antigen se20 9|se20 9|SNF1 sucrose nonfermenting like kinase|STE20 related kinase|SLK_HUMAN.|STE20 like serine/threonine-protein kinase|LOSK
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10257-744
- Supplier no.:BS-11484R-CY7
Specifications
About this item
SLK is a member of the serine/threonine kinase subfamily, Ste20. This subfamily is comprised of several mammalian kinases which exhibit sequence similarity to the Saccharomyces cerevisiae serine/threonine kinase Ste20, a protein involved in relaying signals from G protein-coupled receptors to cytosolic MAP kinase cascades. Members of this subfamily include KHS, GLK, YSK1, HPK1, Krs-1, Krs-2, GC kinase, HGK and SLK. SLK is a ubiquitously expressed protein that localizes to the cytoplasm and contains an N-terminal protein kinase domain, a central coiled-coil domain and a C-terminal ATH domain. SLK is activated through cleavage by caspase-3. SLK indirectly associates with microtubules and plays an important role in cellular stress, cell motility, cell death and cytoskeletal dynamics.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: STK2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat