To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Doublecortin-like Kinase 1
- Antigen Symbol:DCAMKL1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Doublecortin-like and CAM kinase-like 1|Serine/threonine-protein kinase DCAMKL1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10257-064
- Supplier no.:BS-11455R-FITC
Lissencephaly (smooth brain) is an abnormality of brain development characterized by incomplete neuronal migration and a smooth cerebral surface, manifesting as severe mental retardation. Genetic analysis has identified two proteins that are mutated in some cases of lissencephaly, designated lissencephaly-1 protein (LIS1) and doublecortin. LIS1 displays sequence homology to ?subunits of heterotrimeric G proteins, and doublecortin contains a consensus Abl phosphorylation site. In addition, the DCAMKL1 (doublecortin-like and CAM kinase-like 1) protein shows homology to doublecortin. All three proteins are highly expressed in developing brain and may function together to regulate microtubules involved in neuronal migration. The DCAMKL1 protein encodes a functional kinase that is capable of phosphorylating myelin basic protein and itself, but its kinase activity does not appear to affect its microtubule polymerization activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DCAMKL1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat