To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Acetoacetyl-CoA Synthetase
- Antigen Symbol:AACS
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:65985
- Antigen Synonyms:Acyl-CoA synthetase family member 1|Protein sur-5 homolog|Acetoacetyl-CoA synthetase|AACS_HUMAN.|ACSF1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10254-694
- Supplier no.:BS-11383R-CY3
ACSF1 is a 672 amino acid protein belonging to the ATP-dependent AMP-binding enzyme family. Encoded by a gene that maps to human chromosome 12q24.31, ACSF1 is highly expressed in kidney, heart and brain, and shows similar neural expression as HMGCR (3-hydroxy-3-methylglutaryl-CoA reductase). Existing as three alternatively spliced isoforms, ACSF1 participates in ATP binding, ligase activity, acetoacetate-CoA ligase activity and nucleotide binding. The ACSF1 promoter is a known PPAR?target gene, with the nuclear receptor recruited to the ACSF1 promoter by direct interaction with stimulating protein-1 (Sp1). ACSF1 activates acetoacetate and is highly regulated by modulators that affect HMGCR and cholesterol biosynthesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AACS
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat