Specifications
- Antibody Type:Primary
- Antigen Name:Syntaxin 12
- Antigen Symbol:STX12
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:65033
- Antigen Synonyms:MGC51957|STX12|STX14|STX13
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10254-608
- Supplier no.:BS-11378R-FITC
Specifications
About this item
Syntaxin 13 is an integral membrane protein that belongs to the t-SNARE family, a group of proteins involved in protein transport. Confocal immunofluoresence and electron microscopy studies have shown that syntaxin 13 is primarily localized to tubular early and recycling endosomes, where it colocalizes with transferrin receptor, and it is also localized in endosomal vacuoles. Syntaxin 13 has been found to be expressed in all tissues, with higher levels of the protein found in brain, lung, spleen, thymus and testes. Immunoprecipitation studies show that syntaxin 13 complexes with beta-SNAP, VAMP2/3, and SNAP25. The binding of this complex to alpha-SNAP and NSF is terminated in the presence of ATP. These results suggest that syntaxin 13 is a SNARE protein which mediates the recycling protein flow through tubulo vesicular recycling endosomes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: STX12
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat