To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Synaptic Vesicle Glycoprotein 2C
- Antigen Symbol:SV2C
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:22987
- Antigen Synonyms:SV2C_HUMAN.|Synaptic vesicle protein 2c
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10254-344
- Supplier no.:BS-11366R-CY7
Specifications
About this item
In all vertebrates, SV2 proteins are abundant, hydrophobic, membrane glycoproteins that are expressed as two major isoforms, SV2A and SV2B, and one minor isoform, SV2C. SV2 proteins are differentially expressed in the brain and are present on all synaptic vesicles, independent of transmitter type. SV2A is abundantly expressed in the subcortex, specifically in the synaptic vesicles of all presynaptic nerve terminals, and also in most neuroendocrine secretory granules. SV2B displays a more restricted pattern of expression in that it is only present on a small subset of synapses in the hippocampus and cortex. SV2A and SV2B are funtionally redundant and are required for maintaining normal brain function in vertebrates. SV2A and SV2B mediate synaptic transmission by regulating cytoplasmic Ca2+ levels in the nerve terminal during repetitive stimulation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SV2C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat