Specifications
- Antibody Type:Primary
- Antigen Name:Transmembrane emp24 domain-containing protein 10
- Antigen Symbol:TMED10
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:TMED10|Tmp 21 I|21 kDa transmembrane trafficking protein|Transmembrane protein Tmp21|TMED 10|Transmembrane emp24 domain containing protein 10|S31III125|p23|Transmembrane trafficking protein 21kD|S31I125|1110014C03Rik|Transmembrane emp24-like trafficking protein 10 yeast|MGC102351|p24delta|p24delta1|TMEDA_HUMAN.|p24 family protein delta 1|Tmp 21
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10252-586
- Supplier no.:BS-11262R-FITC
Specifications
About this item
MP21 is a member of the EMP24/GP25L/p24 cargo family of proteins that regulates vesicular trafficking in the early secretory pathway. TMP21 is a ubiquitously expressed single-pass type I membrane protein localizing to the Golgi cisternae and the plasma membrane. It contains one GOLD (Golgi dynamics) domain and participates in protein transport and quality control between the endoplasmic reticulum (ER) and the Golgi complex. In addition, TMP21 is a component of the heteromeric secretase complex (or Presenilin complex) and functions to regulate the ?secretase activity. Two isoforms, namely TMP21-I and TMP21-II, exist for this protein.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat