Anti-SOST Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
10363-866
: Bioss
- Antibody Type:Primary
- Antigen Name:Sclerostin
- Antigen Symbol:SOST
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Gene ID:50964
- Antigen Synonyms:VBCH|CDD|Sclerostin|UNQ2976/PRO7455/PRO7476|SOST|SOST1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:150-200/213
- Purification:Purified by Protein A
- Cat. No.:10363-866
- Supplier no.:BS-10200R-FITC
Negative regulator of bone growth that acts through inhibition of Wnt signaling and bone formation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SOST
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat