Anti-Batroxobin Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # 10358-888
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Batroxobin
- Antigen Symbol:Batroxobin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10358-888
- Supplier no.:BS-0912R-A647
Specifications
About this item
No data available.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Batroxobin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species