Anti-SARS putative orflab polyprotein Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10231-864
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Putative Orflab Polyprotein
- Antigen Symbol:POP
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10231-864
- Supplier no.:BS-0132R-HRP
Specifications
About this item
Severe acute respiratory syndrome (SARS) is a viral respiratory illness caused by a coronavirus, called SARS-associated coronavirus (SARS-CoV). Human coronaviruses (HCoVs) were previously only associated with mild diseases. The SARS-CoV genome contains five major open reading frames (ORFs) that encode the replicase polyprotein; the spike (S), envelope (E), and membrane (M) glycoproteins; and the nucleocapsid protein (N).
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: SARS putative orflab polyprotein
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus