Specifications
- Length:140 mm (5½")
- Material:Stainless steel
- Cat. No.:10184-418
- Supplier no.:97-358
- Blade type:Smooth, Blunt/Blunt, Straight
Specifications
About this item
Merit™ Littauer Stitch scissors are used for suture removal.
These littauer scissors have a small hook shaped tip on one blade, that slides under sutures to slightly lift them before cutting for removal. This hook holds the suture, so it does not slip off the blade of the scissors prior to cutting. Littauer Stitch scissors have larger, less delicate tips, and are used for removing larger sutures.
They should not be used for cutting tissue.