ContactUsLinkComponent
Surgi-OR™ Spencer Stitch Scissors, OR Grade, Sklar
Surgi-OR™ Spencer Stitch Scissors, OR Grade, Sklar
Catalog # 89023-588
Supplier:  Sklar
undefined
Surgi-OR™ Spencer Stitch Scissors, OR Grade, Sklar
Catalog # 89023-588
Supplier:  Sklar
Supplier Number:  95-346
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    89 mm (3½")
  • Material:
    Stainless steel
  • Cat. No.:
    89023-588
  • Supplier no.:
    95-346
  • Blade type:
    Smooth, Straight, Blunt/Blunt
Specifications
About this item
Surgi-OR™ Spencer Stitch scissors are used for suture removal.
They have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They have smaller, more delicate tips used for removing smaller sutures. They should not be used for cutting tissue.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.