Anti-Cardiac Troponin, I subunit Goat Polyclonal Antibody
10013-964
- Antibody Type:Primary
- Antigen Name:Cardiac Troponin, I Subunit
- Antigen Symbol:Cardiac Troponin, I Subunit
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Cross Adsorption:No
- Format:Purified IgG
- Form:Clear, colorless liquid, 0.2 µm filtered
- Antigen Synonyms:Purified Human Cardiac Troponin|I subunit (Part# Hu-090-M)
- Storage Buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature:2-8 °C
- Concentration:4-6 mg/ml
- Shipping Temperature:2-8 °C
- Immunogen:Human Cardiac Troponin, I subunit (Part# Hu-090-M)
- Purification:Affinity purified using solid-phase peptide 3 (-RAYATEPHAKKKS-)
- Cat. No.:10013-964
- Supplier no.:IR2915
This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications
Type: Primary
Antigen: Cardiac Troponin, I subunit
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human