Copper, Oil based standard solution, Specpure®, Cu 5000 µg/g, Thermo Scientific
Catalog # AA47225-18
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:5000 µg/g
- Matrix:Oil
- Size:50 g
- MDL No.:MFCD00010965
- Cat. No.:AA47225-18
- Element:Copper
- Supplier no.:47225-18
Specifications
About this item
Copper, Oil based standard solution, Specpure|r, Cu 5000^mg/g
Physical Form: Liquid
MDL No.: MFCD00010965