Cone Style, N95 Particulate Respirator, Tronex®
Supplier: Tronex
Certificates
About this item
Tronex® filtering respiration mask is a NIOSH N95 approved Particulate Respirator
- Conforms to NIOSH 42CFR Part 84 Standard as N95 Class
- 95% filtration efficiency against particulate solids (0.3μ) and aerosols free of oils
- Effectively protects against the transmission of airborne pathogens such as avian and swine influenza, tuberculosis (TB), dust, and pollen
- Meets CDC Guidelines for TB exposure control
- Designed for extended wear with comfortable head straps
- Features malleable nose bridge and foam bridge seal for customized fit
- 100% latex-free product
- Unisize: Engineered for one size fits all
- Small: Engineered for smaller facial structures
It can be used to protect against the inhalation of particulates and airborne pathogens. Available in unisize and small.