Phosphate Buffer, pH 7.2, BD Diagnostics
Supplier: BD
Certificates
About this item
Phosphate Buffer, pH 7.2 is used for the preparation of dilution blanks for use in the examination of waters, dairy products, foods and other materials.
Phosphate buffer is used in the preparation of dilution blanks for use in microbiological testing rather than unbuffered water in order to standardize this potential variable due to the wide variation in the pH of purified water from multiple sources. Sodium carbonate is a pH regulator.
Certifications: Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP)1-3 performance
specifications.
specifications.