ContactUsLinkComponent
AMPIGENE® HS Taq DNA Polymerase, Enzo Life Sciences
AMPIGENE® HS Taq DNA Polymerase, Enzo Life Sciences
Catalog # 10014-682
Supplier:  Enzo Life Sciences
CAS Number:  
AMPIGENE® HS Taq DNA Polymerase, Enzo Life Sciences
Catalog # 10014-682
Supplier:  Enzo Life Sciences
Supplier Number:  PRT101-0500
CAS Number:  
Specifications
  • Description:
    AMPIGENE® HS Taq DNA Polymerase
  • Size:
    500 units
  • Cat. No.:
    10014-682
  • Supplier no.:
    PRT101-0500
Specifications
About this item
Increased sensitivity standard PCR for a broader range of samples, with enhanced speed, yield, and specificity.
For standard PCR applications that require higher sensitivity.
AMPIGENE® HS Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE® HS Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. Hot-start technology is used to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine.The AMPIGENE® HS Taq DNA Polymerase is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine.
Ordering information: Contains AMPIGENE® HS Taq DNA polymerase (5u/µl) 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).