Cellular Senescence Live Cell Analysis Assay, Enzo Life Sciences
Catalog # 10001-774
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Cat. No.:10001-774
- Supplier no.:KIT130-0010
- No. of tests:10
Specifications
About this item
Simple assay to quantify senescence-associated β-galactosidase activity, in live cells, using flow cytometry or fluorescent microscopy.
For the study of cellular senescence in live cells.
Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells.
Ordering information: Contains Cell pretreatment solution (1000x) and SA-?-gal Substrate (200x).