Adipogenesis Assay Kit, Enzo Life Sciences
10014-582
:
- Cat. No.:10014-582
- No. of wells:24
- Supplier no.:KIT103-0005
Convenient method for the enhanced induction and detection of adipogenesis, in the classic 3T3L-1 model.
For the induction and colorimetric detection of adipogenesis.
The Adipogenesis Assay Kit provides all the reagents needed to induce and detect adipogenesis. Optimized Differentiation, Insulin, and Enhancer solutions ensure maximal adipocyte induction. The Adipogenesis Dye allows for visual confirmation of differentiation and can also be easily extracted allowing measurement using a spectrophotometer or microplate reader.
: Contains 1000x Differentiation Solution (Dexamethasone and IBMX), 1000x Insulin Solution, 1000x Enhancer Solution (Rosiglitazone), 1x Cell Fixative, 1x Adipogenesis Dye (Oil Red O), 1x Extraction Solution.