ContactUsLinkComponent
AMPIGENE® DNA Ladder 50 to 1500 bp, Enzo Life Sciences
AMPIGENE® DNA Ladder 50 to 1500 bp, Enzo Life Sciences
Catalog # 10014-496
Supplier:  Enzo Life Sciences
CAS Number:  
AMPIGENE® DNA Ladder 50 to 1500 bp, Enzo Life Sciences
Catalog # 10014-496
Supplier:  Enzo Life Sciences
Supplier Number:  GEN102-0100
CAS Number:  
Specifications
  • Size:
    100 loads
  • Environmentally Preferable:
  • Cat. No.:
    10014-496
  • Supplier no.:
    GEN102-0100
Specifications
About this item
Ready to use DNA standards for quantifying PCR yield
For size determination and DNA quantification using agarose electrophoresis
AMPIGENE® DNA Ladder 50-1,500 bp contains DNA fragments ranging from 50bp to 1,500bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 200bp, 500bp and 1,200bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5µl load, for determining the approximate mass of DNA in samples with comparable intensity and size.
Ordering information: Contains Ready-to-use AMPIGENE® DNA Ladder 50-1,500 bp and Sample Loading Buffer.