Order Entry
Puerto Rico
ContactUsLinkComponent
GFP CERTIFIED® FLUOFORTE® Reagent (Non-AM), Enzo Life Sciences
GFP CERTIFIED® FLUOFORTE® Reagent (Non-AM), Enzo Life Sciences
Catalog # 10014-480
Supplier:  Enzo Life Sciences
CAS Number:  
GFP CERTIFIED® FLUOFORTE® Reagent (Non-AM), Enzo Life Sciences
Catalog # 10014-480
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-52555-0250
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Reagent (Non-AM) GFP-Certified™ FluoForte™
  • Size:
    50 µg
  • Purity:
    ≥95% (HPLC)
  • Molecular Weight:
    ~800
  • Cat. No.:
    10014-480
  • Supplier no.:
    ENZ-52555-0250

Specifications

About this item

Calcium dye.

The GFP CERTIFIED® FLUOFORTE®Reagent (Non-AM) is the most sensitive fluorescent calcium indicator relative to other commercially available red fluorescent dyes. It can be used for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Esterase hydrolysis is not required for live cell analysis via flow cytometry or fluorescence microscopy.