Sentry Urinalysis Controls, Thermo Scientific
Catalog # 10154-278
Supplier: Microgenics Corporation (Mas)
CAS Number:
Specifications
- Description:Sentry Urinalysis Controls, Bi-Level
- Size:4 Vials x 25 mL
- Cat. No.:10154-278
- Supplier no.:URN5005
Specifications
About this item
Verify the performance of various hCG test kits with MAS™ Sentry Urinalysis Controls
Ready-to-use liquid controls require no dilution or reconstitution; Level 1 is an abnormal control; Level 2 is a normal control with hCG and microalbumin; May be used to verify the performance of various hCG test kits.
Analytes: Bilirubin, Blood, Glucose, Human Chorionic Gonadotropin (hCG), Leukocytes, Ketones.
MAS Sentry Urinalysis Controls can be stored for 12 months at room temperature and 18 months at 2 to 8°C. They are ideal for a wide variety of urinalysis strips.
Ordering information: Each pack contains two vials of Level 1 and Level 2 controls (4 total vials).