MAS™ Serum Tox Control Set, Thermo Scientific
Catalog # 10154-280
Supplier: Microgenics Corporation (Mas)
CAS Number:
Specifications
- Description:Serum Tox Control Set
- Size:6 Vials x 5 mL
- Cat. No.:10154-280
- Supplier no.:10011608
Specifications
About this item
Achieve reliable and accurate results from testing with the MAS™ Serum Tox Control Set
Multi-Pack, 3 Levels: 2 bottles of each level, 5mL each level.
Analytes: Multianalyte, Barbiturate (Secobarbital), Benzodiazepine (Diazepam), Tricyclic Antidepressants (Nortriptyline) (for use with DRI Reagents).
This control set is used as a consistent test sample of known concentration for monitoring assay conditions in the semi-quantitative measurement of Benzodiazepine and Barbiturates in human serum and plasma, and Tricyclic Antidepressant drugs (TCA) in human serum, plasma, and urine.