CORIO™ CD Open Heating Bath Circulators, JULABO
About this item
CORIO™ CD heating circulator combinations with transparent or stainless steel bath tanks for internal or external temperature applications from +20 to +100°C (plastic tanks) or +20 to +150°C (SS tanks)
Includes CORIO CD immersion circulator with M16x1 male pump connections, 8 and 12mm hose barb sets and bath tank. Bath tank lid included with B5 tank. Stainless steel tanks from 13 - 39 L include drain tap.
To adapt to your individual application we offer a comprehensive range of accessories.
CORIO CD circulator features a bright LED display with easy user interface. Includes pump connections for external fluid flow. A lever below the display enables simple adjustment between internal and external fluid flow. The USB port facilitates external communication.
Certifications: CE marked.
Ordering information: Includes CORIO CD immersion circulator, bath clamp, 8 and 12 mm hose barbs and bath tank.
5 Options Available Below
Product Details & Documents
CORIO™ CD heating circulator combinations with transparent or stainless steel bath tanks for internal or external temperature applications from +20 to +100°C (plastic tanks) or +20 to +150°C (SS tanks)
Includes CORIO CD immersion circulator with M16x1 male pump connections, 8 and 12mm hose barb sets and bath tank. Bath tank lid included with B5 tank. Stainless steel tanks from 13 - 39 L include drain tap.
To adapt to your individual application we offer a comprehensive range of accessories.
CORIO CD circulator features a bright LED display with easy user interface. Includes pump connections for external fluid flow. A lever below the display enables simple adjustment between internal and external fluid flow. The USB port facilitates external communication.
Certifications: CE marked.
Ordering information: Includes CORIO CD immersion circulator, bath clamp, 8 and 12 mm hose barbs and bath tank.
Documents
10015-726 data sheet 10015-728 data sheet 10015-730 data sheet 10015-732 data 10015-734 data
Datasheet
Julabo CORIO Immersion Circulator
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- 10015-726
- 10015-728
- 10015-732
- 10015-736
- 10015-740
- 10015-744
- Pump Flow Rate:
- 15 L/min; 5.1 psi
- 15 L/min; 5.1 psi
- 15 L/min; 5.1 psi
- 15 L/min; 5.1 psi
- 15 L/min; 5.1 psi
- 15 L/min; 5.1 psi
Specifications
Product Family Options
Product Information
- ModelTemperature rangeBath capacityHeating capacityAvailabilityPrice
- Supplier Number: 9012305.02Specifications:
More
Specifications
Cat. No.10015-726Pump Flow Rate15 L/min; 5.1 psi - Supplier Number: 9012319.02Specifications:
More
Specifications
Cat. No.10015-728Pump Flow Rate15 L/min; 5.1 psi - CORIO™ CD-B5, with stainless steel tank20...150 °C5 L1 kWDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.10015-732Pump Flow Rate15 L/min; 5.1 psi - Supplier Number: 9012417.02Specifications:
More
Specifications
Cat. No.10015-736Pump Flow Rate15 L/min; 5.1 psi - Supplier Number: 9012427.02Specifications:
More
Specifications
Cat. No.10015-740Pump Flow Rate15 L/min; 5.1 psi - Supplier Number: 9012439.02Specifications:
More
Specifications
Cat. No.10015-744Pump Flow Rate15 L/min; 5.1 psi
Accessories
Product Information
- Catalog #DetailsAvailabilityPrice
- USB Interface Cable, 2 m, Type A-BUSB Interface Cable, 2 m, Type A-B10015-820For use with CORIO CDLog in for availabilityEach - $72.39
Product Information
- Catalog #For use withAvailabilityPrice
- Stainless Steel lift-up Gable Bath CoverStainless Steel lift-up Gable Bath Cover10015-856For use with B13, B17Log in for availabilityEach - $1,246.34
Product Information
- Catalog #For use withAvailabilityPrice
- Plastic Test Tube Rack for 21 Tubes, 30 mm dia. (Range 100 °C)Plastic Test Tube Rack for 21 Tubes, 30 mm dia. (Range 100 °C)10015-876For use with B13, B17, B19, B27, BT19, BT27; PURA water bathsLog in for availabilityEach - $194.01
Product Information
- Catalog #For use withAvailabilityPrice
- Plastic Test Tube Rack for 60 Tubes 100×16/17 mm dia. (Range 100 °C)Plastic Test Tube Rack for 60 Tubes 100×16/17 mm dia. (Range 100 °C)10015-872For use with B13, B17, B19, B27, BT19, BT27; PURA water bathsLog in for availabilityEach - $195.29
Product Information
- Catalog #For use withAvailabilityPrice
- Plastic Test Tube Rack for 90 Tubes, 75×12/13 mm dia. (Range 100 °C)Plastic Test Tube Rack for 90 Tubes, 75×12/13 mm dia. (Range 100 °C)10015-874For use with B13, B17, B19, B27, BT19, BT27; PURA water bathsLog in for availabilityEach - $194.01
Product Information
- Catalog #DetailsAvailabilityPrice
- Cooling-coil for counter-cooling with tap waterCooling-coil for counter-cooling with tap water10015-848For use with CORIO C, CDLog in for availabilityEach - $398.86
Product Information
- Catalog #DetailsAvailabilityPrice
- Stainless steel lift-up gable bath coverStainless steel lift-up gable bath cover10015-860For use with B33Log in for availabilityEach - $1,365.82
Product Information
- Catalog #For use withAvailabilityPrice
- Stainless Steel Flat Bath CoverStainless Steel Flat Bath Cover10015-862For use with B13, B17Log in for availabilityEach - $560.22
Product Information
- Catalog #For use withAvailabilityPrice
- Stainless Steel Flat Bath Cover
Product Information
- Catalog #For use withAvailabilityPrice
- Stainless Steel Test Tube Rack, 42 Tubes, 75×12/13 mm dia. (Range 150 °C)Stainless Steel Test Tube Rack, 42 Tubes, 75×12/13 mm dia. (Range 150 °C)10015-880For use with B5, BT5Log in for availabilityEach - $332.26
Product Information
- Catalog #For use withAvailabilityPrice
- Immersion-height Adjustable PlatformImmersion-height Adjustable Platform10015-888For use with B19, B27Log in for availabilityEach - $1,078.00
Product Information
- Catalog #AvailabilityPrice
- Hollow balls, polypropylene, 20 mm dia., Range 100 °C (to reduce heat loss, evaporation, oxygen input, odors)Hollow balls, polypropylene, 20 mm dia., Range 100 °C (to reduce heat loss, evaporation, oxygen input, odors)89422-550Log in for availabilityPack of 1,000 - $432.79