Wet/Tank Blotting System, G-Biosciences
Catalog # 10001-838
Supplier: G-Biosciences
Specifications
- Power:40W
- Cat. No.:10001-838
- Transfer area:95 x 87 mm
- Gel size:9.5×8.7 cm
- Supplier no.:BT301
Specifications
About this item
In high quality acrylic with platinum electrodes, the Wet/Tank Blotting System provides high quality and rapid blotting (electro-transfer) of mini handcast and precast gels
A seamless, injection-molded transparent buffer tank prevents leakage and breakage. Suitable for rapid (one hour) transfers or overnight low-intensity transfers, the color coded cassettes and electrodes ensure easy and correct orientation and error-free set-up.
Latches on the gel holder cassette allow for easy handling, simplified insertion and removal of the gel holder cassette from supporting body for transfer (electrode assembly).
It is designed for the transfer of one or two gels in the same unit.