Order Entry
Puerto Rico
ContactUsLinkComponent
Red 598 dUTP, Enzo Life Sciences
Red 598 dUTP, Enzo Life Sciences
Catalog # 10014-646
Supplier:  Enzo Life Sciences
CAS Number:  
Red 598 dUTP, Enzo Life Sciences
Catalog # 10014-646
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-42855
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Red 598 dUTP
  • Size:
    25 nmol
  • Purity Detail:
    Purified by ion-exchange chromatography
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    1mM
  • Formulation:
    Liquid. Solution in water.
  • Cat. No.:
    10014-646
  • Supplier no.:
    ENZ-42855

Specifications

About this item