Order Entry
Puerto Rico
ContactUsLinkComponent
Aqua 431 dUTP (lyophilized), Enzo Life Sciences
Aqua 431 dUTP (lyophilized), Enzo Life Sciences
Catalog # 10014-644
Supplier:  Enzo Life Sciences
CAS Number:  
Aqua 431 dUTP (lyophilized), Enzo Life Sciences
Catalog # 10014-644
Supplier:  Enzo Life Sciences
Supplier Number:  42853L-0050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Aqua 431 dUTP (Lyophilized)
  • Size:
    50 nmol
  • Purity Detail:
    Purified by ion-exchange chromatography
  • Purity:
    ≥93% (HPLC)
  • Formulation:
    Lyophilized
  • Cat. No.:
    10014-644
  • Supplier no.:
    42853L-0050

Specifications

About this item

0.14