Specifications
- Antibody Type:Primary
- Antigen Name:B-cell receptor-associated protein 29
- Antigen Symbol:BCAP29
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:Center
- Format:IgG
- Form:Liquid (sterile filtered)
- Gene ID:55973
- Antigen Synonyms:BAP29
- Concentration:1 mg/mL by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-BAP29 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 17 amino acid synthetic peptide from near the internal region of human Bap29 Bap29 antibody is specific for isoform a of Bap29.
- Purification:Anti-BAP29 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with BAP29 from other sources has not been determined.
- Cat. No.:10802-000
- Supplier no.:600-401-Z28
Specifications
About this item
Bap29 and the related protein Bap31 are endoplasmic reticulum (ER) and ER-vesicle membrane proteins and members of the B-cell receptor-associated protein family. These two proteins are highly homologous and can form homo- and heterodimers. Both Bap29 and Bap31 interact with membrane-bound immunoglobulins (mIgs), such as IgM and IgD, which with Ig-alpha/Ig-beta heterodimers form B cell antigen receptors. Binding of the Bap29/Bap31 heterodimer correlates with the ER retention of non-Ig-alpha/Ig-beta bound mIg complexes, suggesting that Bap29 and Bap31 may act as chaperones transmembrane regions of various proteins. Bap29 possesses multiple isoforms.
Recommended Dilutions: ELISA: 1:5000-1:20,000; Immunohistochemsitry: 10 µg/mL; Immunofluorescence Microscopy: 20 µg/mL; Western Blot: 0.5 - 2 ?g/mL; contains 0.02% (w/v) Sodium Azide
Type: Primary
Antigen: BCAP29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat