Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TEP1 Rabbit Polyclonal Antibody
Anti-TEP1 Rabbit Polyclonal Antibody
Catalog # 10802-454
undefined
Anti-TEP1 Rabbit Polyclonal Antibody
Catalog # 10802-454
Supplier Number:  600-401-AW9

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Telomerase-associated Protein 1
  • Antigen Symbol:
    TEP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    C-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    7011
  • Antigen Synonyms:
    TLP1|VAULT2|TP1|Telomerase protein 1|Telomerase-associated protein 1|TROVE1|p240|Telomerase protein component 1
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-DOK1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to amino acids near the C-terminus of human DOK1.
  • Purification:
    Anti-DOK1 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with DOK1 from other sources has not been determined.
  • Cat. No.:
    10802-454
  • Supplier no.:
    600-401-AW9

Specifications

About this item

Signals from most growth factors and cytokines are transduced by receptor tyrosine kinases or non-receptor tyrosine kinases. Activated tyrosine kinases phosphorylate their substrates, which mediate the cellular response to extracellular stimuli. A long-sought major substrate termed p62dok (downstream of tyrosine kinase) for many tyrosine kinases including c-kit, v-abl, v-Fps, v-Src, v-Fms, and activated EGF, PDGF, IGF, VEGF and insulin receptors was identified recently from human and mouse by several laboratories. Upon phosphorylation, p62dok forms a complex with the ras GTPase-activating protein (RasGAP). p62dok represents a new family with very recently identified p56dok.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 10 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TEP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human