Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-DDX3X Rabbit Polyclonal Antibody
Anti-DDX3X Rabbit Polyclonal Antibody
Catalog # 10801-120
undefined
Anti-DDX3X Rabbit Polyclonal Antibody
Catalog # 10801-120
Supplier Number:  600-401-AU4

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked
  • Antigen Symbol:
    DDX3X
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Center
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    1654
  • Antigen Synonyms:
    DDX14|DBX|DDX3|HLP2|CAP-Rf
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-DDX3 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 16 amino acid synthetic peptide from near the internal region of human DDX3.
  • Purification:
    Anti-DDX3 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. DDX3 antibody will detect both DDX3 and DBY.
  • Cat. No.:
    10801-120
  • Supplier no.:
    600-401-AU4

Specifications

About this item

DDX3 contains all of the motifs of the DEAD-box family of RNA helicases, including the Asp-Glu-Ala-Asp sequence that gives the protein family its name and distinguishes it from other RNA helicases. DDX3 is localized to the X chromosome and has a highly conserved functional homolog (DBY) on the Y chromosome. DDX3 is thought to be involved in RNA splicing, RNA transport, and translation initiation. It has also been shown to be involved in cell growth control and is deregulated in hepatitis virus-associated hepatocellular carcinoma. Recent experiments suppressing DDX3 expression blocked HIV-1 RNA export from the nucleus, suggesting that DDX3 functions as a shuttling protein that transports the HIV-1 protein Rev and its cofactor CRM1 from the nucleus to the cytoplasm.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 10 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DDX3X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat