Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TNFRSF10C Rabbit Polyclonal Antibody
Anti-TNFRSF10C Rabbit Polyclonal Antibody
  10801-098
undefined
Anti-TNFRSF10C Rabbit Polyclonal Antibody
  10801-098
 :  600-401-AT6

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Tumor Necrosis Factor Receptor Superfamily Member 10C
  • Antigen Symbol:
    TNFRSF10C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Extracellular
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    8794
  • Antigen Synonyms:
    DCR1|TRID|TRAIL-R3|TRAILR3|DCR1-TNFR|CD263|LIT
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-DcR1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to amino acids in the extracellular domain of human DcR1 precursor.
  • Purification:
    Anti-DcR1 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with DcR1 from other sources has not been determined.
  • Cat. No.:
    10801-098
  • Supplier no.:
    600-401-AT6

 

 

Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors. TRAIL/Apo2L is a new member of the TNF family and induces apoptosis of a variety of tumor cell lines. DR4 and DR5 are the recently identified functional receptors for TRAIL. Two decoy receptors for TRAIL have been identified and designated DcR1/TRID/TRAIL-R3/LIT and DcR2/TRAIL-R4/TRUNDD. DcR1 has extracellular TRAIL-binding domain but lacks intracellular signaling domain. It is a glycophospholipid-anchored cell surface protein. DcR1 transcripts are expressed in many normal human tissues but not in most cancer cell lines. Overexpression of DcR1 did not induce apoptosis, but attenuated TRAIL-induced apoptosis.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 10 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TNFRSF10C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Extracellular
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat