Order Entry
United States
ContactUsLinkComponent
 
Anti-C1QTNF7 Rabbit Polyclonal Antibody
  10800-952
undefined
Anti-C1QTNF7 Rabbit Polyclonal Antibody
  10800-952
 :  600-401-AQ6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    C1q And tumor necrosis factor related protein 7
  • Antigen Symbol:
    C1QTNF7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    109323
  • Antigen Synonyms:
    8430425G24Rik|Ctrp7|5530401N20Rik
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-CTRP7 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 15 amino acid synthetic peptide from near the C-terminus of human CTRP7.
  • Purification:
    Anti-CTRP7 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. These proteins are often highly modified post-translationally and migrate in SDS-PAGE at positions other than their predicted size.
  • Cat. No.:
    10800-952
  • Supplier no.:
    600-401-AQ6

 

 

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Like the other members of the adiponectin and CTRP protein family, the mature CTRP7 is secreted and can be found in the organism's circulatory system.

Recommended Dilutions: ELISA: 1:5000-1:20,000; Immunofluorescence Microscopy: 2 ?g/mL; Western Blot: 0.5-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: C1QTNF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat