Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-C1QTNF4 Rabbit Polyclonal Antibody
Anti-C1QTNF4 Rabbit Polyclonal Antibody
Catalog # 10800-940
undefined
Anti-C1QTNF4 Rabbit Polyclonal Antibody
Catalog # 10800-940
Supplier Number:  600-401-AQ0

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    C1q And tumor necrosis factor related protein 4
  • Antigen Symbol:
    C1QTNF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    C-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    114900
  • Antigen Synonyms:
    CTRP4|ZACRP4
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-CTRP4 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 16 amino acid synthetic peptide from near the C-terminus of human CTRP4.
  • Purification:
    Anti-CTRP4 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. These proteins are often highly modified post-translationally and migrate in SDS-PAGE at positions other than their predicted size.
  • Cat. No.:
    10800-940
  • Supplier no.:
    600-401-AQ0

Specifications

About this item

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of mouse CTRP4 have been reported.

Recommended Dilutions: ELISA: 1:10,000-1:40,000; Immunohistochemsitry: 10 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-4 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: C1QTNF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat