Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CASP6 Rabbit Polyclonal Antibody
  10802-280
undefined
Anti-CASP6 Rabbit Polyclonal Antibody
  10802-280
 :  600-401-AD7

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Caspase 6
  • Antigen Symbol:
    CASP6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Center
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    839
  • Antigen Synonyms:
    caspase 6|apoptotic protease MCH-2|caspase-6|CASP-6|apoptosis-related cysteine protease
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-Caspase-6 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 15 amino acid synthetic peptide from near the internal region of human Caspase-6.
  • Purification:
    Anti-Caspase-6 Antibody is immunoaffinity purified. Depending on cell lines or tissues used, either full-length or other cleavage products may be observed.
  • Cat. No.:
    10802-280
  • Supplier no.:
    600-401-AD7

 

 

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. The apoptotic subfamily can be further divided into initiator caspases, which are activated in response to death signals, and executioner caspases, which are activated by the initiator caspases and are responsible for cleavage of cellular substrates that ultimately lead to cell death. Caspase-6 is an executioner caspase that was identified based on its homology with human caspases 2 and 3 as well as the C. elegans cell death protein CED-3. It possesses two isoforms, of which only the longer form possesses protease activity. Caspase-6 is highly expressed in adult brain and may play a role in several neuronal pathologies.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: 10 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG1
Reactivity: Human