Order Entry
United States
ContactUsLinkComponent
 
Anti-TBC1D10C Rabbit Polyclonal Antibody
Catalog # 10800-520
undefined
Anti-TBC1D10C Rabbit Polyclonal Antibody
Catalog # 10800-520
Supplier Number:  600-401-AB3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TBC1 Domain Family, Member 10C
  • Antigen Symbol:
    TBC1D10C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    374403
  • Antigen Synonyms:
    Carabin|CARABIN|EPI64C|TBC1 domain family member 10C
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-Carabin antibody was prepared from whole rabbit serum produced by repeated immunizations with a 17 amino acid synthetic peptide from near the N-terminus of human Carabin.
  • Purification:
    Anti-Carabin Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with Carabin from other sources has not been determined.
  • Cat. No.:
    10800-520
  • Supplier no.:
    600-401-AB3

Specifications

About this item

Antigen binding by the T-cell receptor (TCR) is one of the critical first steps in the immune response, triggering a cascade of signaling pathways that ultimately lead to T-cell activation. Screening a yeast two-hybrid screen of a human T-cell cDNA library with calcineurin, a protein phosphatase involved in multiple signaling pathways including T-cell activation, resulted in the identification of Carabin, a member of the TBC1 domain family of proteins, as a calcineurin-binding protein. Unlike other members of the TBC1 domain protein family which are thought to have a role in regulating cell growth and differentiation, further experiments demonstrated that Carabin is part of a negative regulatory loop for the intracellular TCR signaling pathway as well as an inhibitor of the Ras signaling pathway, suggesting that Carabin may also mediate crosstalk between calcineurin and Ras. Carabin antibody does not recognize TBC1D10A or TBC1D10B. Carabin is known to exist in multiple isoforms.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 2.5 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TBC1D10C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse