Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-DFFB Rabbit Polyclonal Antibody
Anti-DFFB Rabbit Polyclonal Antibody
Catalog # 10802-074
undefined
Anti-DFFB Rabbit Polyclonal Antibody
Catalog # 10802-074
Supplier Number:  600-401-AA5

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    DNA fragmentation factor, beta subunit
  • Antigen Symbol:
    DFFB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    C-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    13368
  • Antigen Synonyms:
    5730477D02Rik|CPAN|Didff|DFF40|CAD|40kDa
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-CAD antibody was prepared from whole rabbit serum produced by repeated immunizations with a 16 amino acid peptide near the C-terminus of murine CAD.
  • Purification:
    Anti-CAD Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with CAD from other sources has not been determined.
  • Cat. No.:
    10802-074
  • Supplier no.:
    600-401-AA5

Specifications

About this item

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A mouse DNase that causes DNA fragmentation was identified recently and designated CAD (for caspase activated deoxyribonuclease). The human homologue of mouse CAD was more recently identified by two groups independently and termed CPAN and DFF40. Human DFF45 and its mouse homologue ICAD are the inhibitors of CPAN/DFF40 and CAD, respectively. Upon cleavage of DFF45/ICAD by activated caspase, DFF40/CAD is released and activated and eventually causes the degradation of DNA in the nuclei. Activation of CAD/DFF40, which causes DNA degradation, is the hallmark of apoptotic cell death.

Recommended Dilutions: ELISA: 1:5,000; Immunohistochemsitry: 1 ?g/mL; Immunofluorescence Microscopy: 10 ?g/mL; Western Blot: 0.5 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DFFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse