Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GLB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-GLB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  10800-326
Anti-GLB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  10800-326
 :  200-4336-0100

 

  • Antibody Type:
    Tag
  • Antigen Name:
    Galactosidase Beta 1
  • Antigen Symbol:
    GLB1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Format:
    IgG
  • Form:
    Lyophilized
  • Antigen Synonyms:
    PPCA|PPGB|RP3-337O18.1|NGBE|GLB2|Beta-Gal|GSL
  • Concentration:
    1.0 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Ambient
  • Immunogen:
    Beta Galactosidase (E.coli)
  • Purification:
    Beta Galactosidase is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.  Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, and anti-Rabbit Serum as well as purified and partially purified Beta Galactosidase [E.coli].  Cross reactivity against Beta Galactosidase from other tissues and species may occur but have not been specifically determined.  Very low background staining has been reported in various assays.
  • Cat. No.:
    10800-326
  • Supplier no.:
    200-4336-0100

 

 

β-galactosidase, (beta-gal or β-gal), is a hydrolase enzyme that catalyzes the hydrolysis of β-galactosides into monosaccharides. It may also cleave fucosides and arabinosides but with much lower efficiency. It is an essential enzyme in the human body. Deficiencies in the protein can result in galactosialidosis or Morquio B syndrome.

Recommended Dilutions: ELISA: 1:2,000 - 1:5,000; Immunohistochemsitry: 1:250; Western Blot: 1:500 - 1:2,000; 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide!

Type: Tag
Antigen: beta Galactosidase
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: