Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-BCL2L14 Rabbit Polyclonal Antibody
Anti-BCL2L14 Rabbit Polyclonal Antibody
Catalog # 10802-040
undefined
Anti-BCL2L14 Rabbit Polyclonal Antibody
Catalog # 10802-040
Supplier Number:  200-401-Z48

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    BCL2-Like 14
  • Antigen Symbol:
    BCL2L14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    N-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    79370
  • Antigen Synonyms:
    Apoptosis facilitator Bcl-2-like protein 14|Apoptosis regulator Bcl-G|Bcl2-L-14|Bcl-G|BCLG
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-Bcl-G antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to 15 amino acids near the N-terminus of human Bcl-G.
  • Purification:
    Anti-Bcl-G Antibody is purified by ion exchange chromatography. Although antibody should react with both isoforms, only the Bcl-GS protein has been observed
  • Cat. No.:
    10802-040
  • Supplier no.:
    200-401-Z48

Specifications

About this item

Members in the Bcl-2 family are critical regulators of apoptosis by either inhibiting or promoting cell death. Bcl-2 homology 3 (BH3) domain is a potent death domain. BH3 domain containing pro-apoptotic proteins, including Bad, Bax, Bid, Bik, and Hrk, form a growing subclass of the Bcl-2 family. A novel BH3 domain containing protein was recently identified and designated Bcl-G. The mRNA of Bcl-G encodes 2 isoforms, Bcl-GL, which is widely expressed in multiple tissues, and Bcl-GS, which is only found in testis. The Bcl-GS protein is predominantly localized to cytoplasmic organelles whereas Bcl-GL was distributed throughout the cytosol. Overexpression of either protein induced apoptosis, although Bcl-GS was far more potent than Bcl-GS. Apoptosis induction was dependent on the BH3 domain and could be suppressed by co-expression with the anti-apoptotic Bcl-XL protein.

Recommended Dilutions: ELISA: 1:25,000-1:100,000; Immunohistochemsitry: 2 µg/mL; Immunofluorescence Microscopy: 10 µg/mL; Western Blot: 2.5 - 5 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCL2L14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat