Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-BCL2 Rabbit Polyclonal Antibody
Anti-BCL2 Rabbit Polyclonal Antibody
Catalog # 10800-300
undefined
Anti-BCL2 Rabbit Polyclonal Antibody
Catalog # 10800-300
Supplier Number:  200-401-Z43

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    B-cell lymphoma 2
  • Antigen Symbol:
    BCL2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    N-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    596
  • Antigen Synonyms:
    PPP1R50|Bcl-2
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-Bcl-2 antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to 15 amino acids near the N terminus of human Bcl-2.
  • Purification:
    Anti-Bcl-2 Antibody is purified by ion exchange chromatography. Cross reactivity with Bcl-2 from other sources has not been determined.
  • Cat. No.:
    10800-300
  • Supplier no.:
    200-401-Z43

Specifications

About this item

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Bcl-2 is the founding member of a family of over 20 proteins that are critical regulators of apoptosis. These can be divided into two classes: those that inhibit apoptosis and those that promote cell death. Bcl-2 is an inner mitochondrial membrane protein that inhibits apoptosis. It is thought to act by interacting with pro-apoptotic Bcl-2 family members such as Bak and Bad. Overexpression of Bcl-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.

Recommended Dilutions: ELISA: 1:10,000-1:40,000; Immunohistochemsitry: 2 µg/mL; Immunofluorescence Microscopy: 10 µg/mL; Western Blot: 1 - 4 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse