Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-X-C Motif) Receptor 4
- Antigen Symbol:CXCR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Format:IgG
- Form:Liquid (sterile filtered)
- Gene ID:7852
- Antigen Synonyms:HM89|LAP-3|WHIM|NPYR|NPYRL|D2S201E|NPY3R|NPYY3R|LCR1|HSY3RR|CD184|WHIMS|LAP3|LESTR|FB22
- Concentration:1 mg/mL by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-CXCR4-Lo antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to nine amino acids near the N-terminus of human CXCR4 isoform a.
- Purification:Anti-CXCR4-Lo Antibody is purified via Protein A column purification. This antibody is specific for the longer isoform of CXCR4.
- Cat. No.:10801-038
- Supplier no.:200-401-AR5
Specifications
About this item
Human immunodeficiency virus (HIV) and related viruses require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, CCR2b and CCR8 in the chemokine receptor family, and four new human molecules GPR15, STRL33, GPR1 and V28 were recently identified as HIV coreceptors. Among them, CXCR4 is a principal coreceptor for T-cell tropic strains of HIV-1 fusion and entry of human white blood cells. CXCR4 is also required for the infection by dual-tropic strains of HIV-1 and mediates CD-4 independent infection by HIV-2. The alpha-chemokine SDF-1 is the ligand for CXCR4 and prevents infection by T-tropic HIV-1. CXCR4 associates with the surface CD4-gp120 complex before HIV enters target cells. CXCR4 messenger RNA levels correlated with HIV-1 permissiveness in diverse human cell types. Antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV binding sites.
Recommended Dilutions: ELISA: 1:100,000; Immunofluorescence Microscopy: 4 ?g/mL; Western Blot: 10 ?g/mL; contains 0.02% (w/v) Sodium Azide
Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: