Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ATF4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: S360A-24]
Anti-ATF4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: S360A-24]
Catalog # 10800-170
Anti-ATF4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: S360A-24]
Catalog # 10800-170
Supplier Number:  200-302-W61
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Activating Transcription Factor 4
  • Antigen Symbol:
    ATF4
  • Clonality:
    Monoclonal
  • Clone:
    S360A-24
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    C-Terminal
  • Format:
    IgG2a
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    468
  • Antigen Synonyms:
    DNA-binding protein TAXREB67|cAMP-dependent transcription factor ATF-4|Cyclic AMP-dependent transcription factor ATF-4|TXREB|TaxREB67|Tax-responsive enhancer element-binding protein 67|CREB2|Activating transcription factor 4|cAMP-responsive element-binding protein 2|CREB-2|Cyclic AMP-responsive element-binding protein 2|ATF4
  • Concentration:
    1mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Wet Ice
  • Immunogen:
    Anti-ATF4 Antibody was produced by repeated immunization of mice with a fusion protein containing amino acids 25-327 of human ATF4.
  • Purification:
    Anti-ATF4 Antibody was purified from concentrated tissue culture supernate by Protein G chromatography. BLAST analysis suggests that it is 85% identical to mouse, 86% identical to rat and <50% identical to ATF5. Will not cross react with ATF5.
  • Cat. No.:
    10800-170
  • Supplier no.:
    200-302-W61

Specifications

About this item

Cyclic AMP-dependent transcription factor ATF-4 (ATF4) is a basic leucine-zipper (bZip) transcription factor, which regulates amino acid metabolism, DNA damage repair, chromatin remodeling, and apoptosis in response to cellular and ER stress. ATF4 works with various proteins, such as C/EBP homology protein (CHOP), aspargine synthetase (ASNS), and cAMP response element (CRE) among others to mediate cellular stress. ATF4 also regulates glucose homeostasis by suppressing beta-cell proliferation and insulin production. Furthermore, ATF4 targets the histone demethylase JMJD3 to alter chromatin structure and enhance gene transcription in response to amino acid deprivation. Anti-ATF4 is ideal for researchers interested in Cell Signaling, Oncology, Cell Differentiation, and Apoptosis; relevant pathways include MAPK signaling pathways, Activation of cAMP-Dependent PKA, CREB pathways, GPCR pathways and Rho Family GTPases.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: User Optimized; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: ATF4
Clonality: Monoclonal
Clone: S360A-24
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: C-Terminal
Host: Mouse
Isotype:
Reactivity: Human, Rat