Order Entry
Puerto Rico
ContactUsLinkComponent
 
TAT Broth, Hardy Diagnostics
  10753-958
Product available on GSA Advantage®
 :  
undefined
TAT Broth, Hardy Diagnostics
  10753-958
 :  D077
Product available on GSA Advantage®
 :  

 

  • Pack type:
    Vial, hinged, 145 ml
  • Description:
    TAT Broth
  • Size:
    50/pack
  • Cat. No.:
    10753-958
  • Volume:
    90 mL
  • Supplier no.:
    D077

 

 

TAT broth is used for the detection of microorganisms from topical drugs, cosmetics, and other viscous materials.