ContactUsLinkComponent
Instru-Guard One-Step Cleaner and Lubricant, Sklar
Instru-Guard One-Step Cleaner and Lubricant, Sklar
Catalog # 89022-276
Supplier:  Sklar
undefined
Instru-Guard One-Step Cleaner and Lubricant, Sklar
Catalog # 89022-276
Supplier:  Sklar
Supplier Number:  10-2800
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Liquid Chemical Cleaner/Lubricant
  • Size:
    3.8 L (1 gal.)
  • Cat. No.:
    89022-276
  • Supplier no.:
    10-2800
Specifications
About this item
Single step cleaner and lubricant for manual washers, decontaminators, or sterilzers
Material composition consists of Potassium Hydroxide, Sodium Metasilicate, and Methanol.