Order Entry
Puerto Rico
ContactUsLinkComponent
Instrument Polish, Sklar
Instrument Polish, Sklar
Catalog # 89022-236
Supplier:  Sklar
undefined
Instrument Polish, Sklar
Catalog # 89022-236
Supplier:  Sklar
Supplier Number:  10-1629

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Cleaner/Lubricant Polish
  • Size:
    236.5 mL (8 oz.)
  • Cat. No.:
    89022-236
  • Supplier no.:
    10-1629

Specifications

About this item

Sklar instrument polish is designed to remove corrosive stains.