Orders LinkContactUsLinkComponent
Disposable Inoculation Needle, Plastic
Disposable Inoculation Needle, Plastic
Disposable Inoculation Needle, Plastic
Supplier:  Copan Diagnostics
Certificates
Disposable Inoculation Needle, Plastic
Disposable Inoculation Needle, Plastic
2 Options Available Below
Product Details & Documents
COPAN Plastic Inoculation Needles have an ultra-smooth surface to avoid gouging agar
Plastic needles are free of lubricants, oils, and electrostatic charges.
Documents
Product Family Options
Product Information
  • Color
    Packaging
    Availability
    Price
  • 30002-172
    Inoculating needle
    Inoculating needle
    Violet
    25/Ziplock Bag
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $397.84
    $397.84
    Case of 1,000
       $397.84
    Supplier Number: COP-NED
  • 30002-108
    Inoculating Needle
    Inoculating Needle
    Violet
    20/Peel Pouch
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $309.02
    $309.02
    Case of 1,000
       $309.02
    Supplier Number: 176CS20