Semiquantitative Standard 1, Specpure®, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Molecular Formula: Matrix: 40% Aqua Regia
Physical Form: Liquid
UN No.: 3093
MDL No.: MFCD02091742
Specifications
- MDL No.:MFCD02091742