Order Entry
Puerto Rico
ContactUsLinkComponent
Cobalt, Oil based standard solution, Specpure®, Co 1000 mg/g, Thermo Scientific
Cobalt, Oil based standard solution, Specpure®, Co 1000 mg/g, Thermo Scientific
Catalog # AA47143-18
CAS Number:  
Cobalt, Oil based standard solution, Specpure®, Co 1000 mg/g, Thermo Scientific
Catalog # AA47143-18
Supplier Number:  047143-18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Oil
  • Size:
    50 g
  • MDL No.:
    MFCD00010935
  • Cat. No.:
    AA47143-18
  • Element:
    Cobalt
  • Supplier no.:
    47143-18

Specifications

About this item

Liquid

Physical Form: Liquid
MDL No.: MFCD00010935