Order Entry
Puerto Rico
ContactUsLinkComponent
Palladium ≥99.9% (metals basis), wire Ø 0.02 mm (0.0008 in)
Palladium ≥99.9% (metals basis), wire Ø 0.02 mm (0.0008 in)
Catalog # AA47134-G1
CAS Number:  
Palladium ≥99.9% (metals basis), wire Ø 0.02 mm (0.0008 in)
Catalog # AA47134-G1
Supplier Number:  047134-G1
CAS Number:  

Specifications

  • Size:
    1 m
  • Cat. No.:
    AA47134-G1
  • Supplier no.:
    47134-G1

Specifications

Specification Test Results

  • Assay Minimum:
    99.9

Specification Test Results

About this item

 :
Formula:Pd
MDL Number:MFCD00011167
 :106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
CAS Number:7440-05-3