Cyanide, Quantofix® Test Strips, Thermo Scientific
Catalog # AA23273-QA
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:1 kit
- MDL No.:MFCD00146594
- UN No.:1993
- Cat. No.:AA23273-QA
- Range:0 - 1 - 3 - 10 - 30 mg/L (ppm) CN⁻
- Supplier no.:23273-QA
Specifications
About this item
The cyanide test is a rapid semi-quantitative exploratory test for cyanide ions in effluent, e.g
UN No.: 1993
MDL No.: MFCD00146594
from electroplating works. It can be used directly to monitor cyanide in waste treatment plants. Only readily dissociable cyanides are detected.
Delivery: Supplied with reagents & strips to measure 0-30ppm cyanide ion in aqueous solution. Comes with 7 grams of sulfate/phosphate buffer powder in one bottle and 25 mL of a pyridine & chloramine-T aq. soln. in an another bottle.