Order Entry
Puerto Rico
ContactUsLinkComponent
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98%
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98%
Catalog # AAJ67344-LB0
CAS Number:  
7-Oxo-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinoline-3-carboxylic acid ≥98%
Catalog # AAJ67344-LB0
Supplier Number:  J67344-LB0
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 mg
  • Cat. No.:
    AAJ67344-LB0
  • Supplier no.:
    J67344-LB0

Specifications

Specification Test Results

  • Assay Minimum:
    98

Specification Test Results

About this item

 :
Formula:C₁₉H₉N₂O₃.C₂H₄O₂
MDL Number:MFCD06411456
 :314.29 g/mol
Storage Temperature:Ambient
CAS Number:52029-86-4
Formula:C₁₉H₁₀N₂O₃
MDL Number:MFCD30685721