To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:0.5 mL
- Storage Temperature:2 - 8 °C
- Cat. No.:AAJ67051-AMI
- Format:Liquid
- Supplier no.:J67051-AMI
Specifications
About this item
This DNA Molecular Weight Marker is used for rapid and precise sizing of DNA strands.
The bands appear in 100 bp increments from 100 to 1,000 bp. The bands are in 1,000 bp increments from 1 to 12 kb. Bands at 1 kb and 5 kb stain brighter to aid identification.